missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ STAT2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30476
This item is not returnable.
View return policy
Description
STAT2 Polyclonal antibody specifically detects STAT2 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| STAT2 | |
| Polyclonal | |
| Tris-buffered Saline 0.1% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| interferon alpha induced transcriptional activator, ISGF-3, MGC59816, p113, signal transducer and activator of transcription 2, signal transducer and activator of transcription 2, 113kD, signal transducer and activator of transcription 2, 113kDa, STAT113 | |
| between 725 and 775 | |
| 20 μg | |
| Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| 6773 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction