Learn More
STAT2 Antibody (S01-2F6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-20074-100ul
 View more versions of this product
Product Code. 18621834
  • € 680.00 / 100µL
Description
STAT2 Monoclonal antibody specifically detects STAT2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Sandwich ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
STAT2
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Mouse
Purified
Western Blot, ELISA, Sandwich ELISA
S01-2F6
Unconjugated
interferon alpha induced transcriptional activator, ISGF-3, MGC59816, p113, signal transducer and activator of transcription 2, signal transducer and activator of transcription 2, 113kD, signal transducer and activator of transcription 2, 113kDa, STAT113
A synthetic peptide of mouse STAT2 (Uniprot # Q9WVL2)
100 μg
Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
6773
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.