missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT2 Antibody (STAT2/2650), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08587AF647
This item is not returnable.
View return policy
Description
STAT2 Monoclonal antibody specifically detects STAT2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| STAT2 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| interferon alpha induced transcriptional activator, ISGF-3, MGC59816, p113, signal transducer and activator of transcription 2, signal transducer and activator of transcription 2, 113kD, signal transducer and activator of transcription 2, 113kDa, STAT113 | |
| Recombinant full-length human STAT2 protein (Uniprot: P52630) | |
| 100 μg | |
| Cancer, Signal Transduction, Transcription Factors and Regulators | |
| 6773 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| STAT2/2650 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction