missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT2, p Tyr689 Antibody (1021D), Alexa Fluor™ 700, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB2890N
This item is not returnable.
View return policy
Description
STAT2 Monoclonal antibody specifically detects STAT2 in Human samples. It is validated for Western Blot, Western Blot, Flow Cytometry
Specifications
| STAT2 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry | |
| 1021D | |
| Western Blot, Simple Western, Flow Cytometry | |
| interferon alpha induced transcriptional activator, ISGF-3, MGC59816, p113, signal transducer and activator of transcription 2, signal transducer and activator of transcription 2, 113kD, signal transducer and activator of transcription 2, 113kDa, STAT113 | |
| Phosphopeptide containing the human STAT2 Y690 site | |
| 0.1 mL | |
| Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| 6773 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction