missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT2 Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB30553.100uL
This item is not returnable.
View return policy
Description
Rabbit polyclonal antibody raised against recombinant human STAT2.
- High specificity
- High reactivity
Specifications
| STAT2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant human STAT2. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| ISGF-3/MGC59816/P113/STAT113 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 6773 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:500), Immunofluorescence (1-4 ug/mL), The optimal working dilution should be determined by the end user. | |
| P52630 | |
| STAT2 | |
| Recombinant protein corresponding to human STAT2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction