missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT3 Antibody (232209) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1799JF585
This item is not returnable.
View return policy
Description
STAT3 Monoclonal antibody specifically detects STAT3 in Human,Mouse,Rat samples. It is validated for Immunoprecipitation,Western Blot,Flow Cytometry (Intracellular Staining),Immunocytochemistry,Gene Knock-Out
Specifications
| STAT3 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| Acute-phase response factor, APRFMGC16063, DNA-binding protein APRF, FLJ20882, HIES, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3 (acute-phase responsefactor) | |
| E. coli-derived recombinant human STAT3, Met1-Asn175, Accession # P40763 | |
| 0.1 mL | |
| Cellular Markers, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| 6774 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Immunoprecipitation, Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Gene Knock-Out | |
| 232209 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction