missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT3 Antibody (OTI21E7), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74378B
This item is not returnable.
View return policy
Description
STAT3 Monoclonal specifically detects STAT3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated.
Specifications
| STAT3 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| STAT3 | |
| Recombinant protein expressed in E.coli corresponding to amino acids 154-175 of human STAT3. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunoassay | |
| OTI21E7 | |
| Western Blot, Immunohistochemistry, Knockout Validated | |
| Acute-phase response factor, APRFMGC16063, DNA-binding protein APRF, FLJ20882, HIES, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3 (acute-phase responsefactor) | |
| Mouse | |
| Affinity Purified | |
| Cellular Markers, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| 6774.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction