Learn More
STAT3 Mouse anti-Human, CoraLite 594, Clone: 3G2D12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-60199-100UL
Description
The STAT3 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human STAT3. This antibody recognizes human, mouse, rat antigen. The STAT3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| STAT3 | |
| Monoclonal | |
| 2000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Immunofluorescence | |
| 3G2D12 | |
| CoraLite 594 | |
| 100 μL | |
| 6774 | |
| IgG2a |