Learn More
STAT3 Mouse anti-Human, CoraLite 594, Clone: 3G2D12, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL594-60199-100UL
Product Code. 16860896
  • € 450.00 / 100µL
Description
The STAT3 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human STAT3. This antibody recognizes human, mouse, rat antigen. The STAT3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
Specifications
STAT3
Monoclonal
2000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Immunofluorescence
3G2D12
CoraLite 594
100 μL
6774