Learn More
STAT3, p Tyr705 Antibody (1004G), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals FAB46071P
Product Code. 18687253
  • € 682.00 / 0.10mL
Description
STAT3 Monoclonal antibody specifically detects STAT3 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
STAT3
Monoclonal
PE
PBS
Rabbit
Protein A or G purified from cell culture supernatant
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry (Intracellular Staining)
1004G
Western Blot, Intracellular Staining by Flow Cytometry
Acute-phase response factor, APRFMGC16063, DNA-binding protein APRF, FLJ20882, HIES, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3 (acute-phase responsefactor)
Phosphopeptide containing the human STAT3 Y705 site, Accession # P40763
0.1 mL
Cellular Markers, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
6774
Store at 4°C in the dark.
IgG2b