missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stat3 (pY705) Mouse, PE, Clone: 4/P-STAT3, BD
Mouse Monoclonal Antibody
€ 535.00 - € 1965.00
Specifications
Antigen | Stat3 (pY705) |
---|---|
Clone | 4/P-STAT3 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15257219
|
BD Biosciences
612569 |
50 Tests |
€ 535.00
Each |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15851619
|
BD Biosciences
562072 |
250 Tests |
€ 1965.00
Each |
Estimated Shipment: 06-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
STAT (Signal transducer and activator of transcription) of Y705 in Stat3 occurs in response to growth factors and cytokines, and is essential for normal transcription activity.
The 4/P-STAT3 monoclonal antibody recognizes the phosphorylated Y705 of Stat3.
Specifications
Stat3 (pY705) | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
4/P-STAT3 | |
Monoclonal | |
Mouse | |
Human, Mouse | |
IgG2a κ | |
Affinity Purified | |
Acute-phase response factor, APRF |
For Research Use Only.