missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT4 Antibody (OTI2F3), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74389IR
This item is not returnable.
View return policy
Description
STAT4 Monoclonal specifically detects STAT4 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| STAT4 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| STAT4 | |
| Full length recombinant protein of human STAT4 (NP_003142) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI2F3 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| signal transducer and activator of transcription 4, SLEB11 | |
| Mouse | |
| Affinity Purified | |
| Cancer, Phospho Specific, Regulatory Immunology, Signal Transduction | |
| 6775.0 | |
| Store at 4°C in the dark. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion