Learn More
STAT4 Monoclonal Antibody (OTI 4H4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA503023
Encompass_Preferred
 View more versions of this product
Product Code. 17508331
  • POA / 100µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
The STAT 4 protein is a transcription factor activated by hormone and cytokines. STAT 4 is most similar to STAT 1 (52%) and STAT 3 (47%). Functionally, STAT 4 is similar to other STAT family members in that it can be tyrosine phosphorylated by JAK 1 or JAK 2. STAT 4 forms homodimers and heterodimers with related STAT family members. Tyrosine phosphorylated STAT 4 can bind the interferon g activated site (GAS), and serine phosphorylation is also required for maximal transcriptional activity. STAT 4 expression is restricted to the thymus, spleen, and testis.
Specifications
Specifications
STAT4
Monoclonal
0.35 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q14765
STAT4
Full length human recombit protein of human STAT4 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI 4H4
Unconjugated
STAT4
SLEB11
Mouse
Affinity Chromatography
RUO
6775
-20° C, Avoid Freeze/Thaw Cycles