missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STAT5 alpha Recombinant Rabbit Monoclonal Antibody (BLR124H)

Rabbit Recombinant Monoclonal Antibody
€ 156.00 - € 474.00
Specifications
| Antigen | STAT5 alpha |
|---|---|
| Clone | BLR124H |
| Concentration | 100 μg/mL |
| Content And Storage | 4°C |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17735809
![]() |
Invitrogen™
MA544437 |
100 μL |
€ 474.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17745809
|
Invitrogen™
MA544438 |
10 μL |
€ 156.00
10µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.
STAT5 alpha is a member of the STAT family of transcription factors. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. STAT5 alpha is activated by, and mediates the responses of many cell ligands, such as IL2, IL3, IL7 GM-CSF, erythropoietin, thrombopoietin, and different growth hormones. Activation of STAT5 alpha in myeloma and lymphoma associated with a TEL/JAK2 gene fusion is independent of cell stimulus and has been shown to be essential for the tumorigenesis. The mouse counterpart of STAT5 alpha is found to induce the expression of BCL2L1/BCL-X(L), which suggests the antiapoptotic function of this protein in cells. STAT5 alpha, along with STAT5 beta, were identified in mouse. The amino acid sequences of STAT5 alpha and STAT5 beta show 96% sequence similarity, and both proteins are co expressed in most tissues in virgin and lactating mice. However, differential accumulation of STAT5 alpha and STAT5 beta mRNA has been reported for both muscle and mammary tissue. STAT5 alpha is critically involved in a variety of physiological functions, including reproduction, lactation, immune function and somatic growth.Specifications
| STAT5 alpha | |
| 100 μg/mL | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2 | |
| P42229, P42230 | |
| 20850, 6776 | |
| Peptide representing a region between residue 744 and the C-terminus (residue 794) of Human signal transducer and activator of transcription 5a. | |
| Antibody |
| BLR124H | |
| 4°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| STAT5A | |
| AA959963; mammary gland factor; mammary gland factor STAT5A; mammary gland factor; transcription factor; Mgf; Mpf; signal transducer and activator of transcription 5; signal transducer and activator of transcription 5a; signal transducer and activator of transcription factor 5A; STAT; STAT5; STAT5A; STAT5A, Mammary Gland Factor; STAT5A/MGF; STAT5B; STATA5 | |
| STAT5A | |
| Primary | |
| Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title