missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STAT5 beta Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119418
This item is not returnable.
View return policy
Description
STAT5 beta Polyclonal Antibody for Western Blot, ICC/IF
The protein encoded by this gene is a member of the STAT family of transcription factors. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. This protein mediates the signal transduction triggered by various cell ligands, such as IL2, IL4, CSF1, and different growth hormones. It has been shown to be involved in diverse biological processes, such as TCR signaling, apoptosis, adult mammary gland development, and sexual dimorphism of liver gene expression. This gene was found to fuse to retinoic acid receptor-alpha (RARA) gene in a small subset of acute promyelocytic leukemias (APLL). The dysregulation of the signaling pathways mediated by this protein may be the cause of the APLL.
Specifications
| STAT5 beta | |
| Polyclonal | |
| Unconjugated | |
| Stat5b | |
| LOC101122961; mammary gland factor STAT5B; Signal transducer and activator of transcription 5a; signal transducer and activator of transcription 5B; STA5B; STAT; STAT5; STAT5B; transcription factor STAT5B | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 6777 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P51692 | |
| Stat5b | |
| Recombinant protein Signal Transducer And Activator Of Transcription 5B. The antigen corresponds to amino acid range 497-749 of the target protein. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction