missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STAT5 beta Recombinant Rabbit Monoclonal Antibody (BLR146J)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544479
This item is not returnable.
View return policy
Description
Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.
The protein encoded by this gene is a member of the STAT family of transcription factors. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. This protein mediates the signal transduction triggered by various cell ligands, such as IL2, IL4, CSF1, and different growth hormones. It has been shown to be involved in diverse biological processes, such as TCR signaling, apoptosis, adult mammary gland development, and sexual dimorphism of liver gene expression. This gene was found to fuse to retinoic acid receptor-alpha (RARA) gene in a small subset of acute promyelocytic leukemias (APLL). The dysregulation of the signaling pathways mediated by this protein may be the cause of the APLL.
Specifications
| STAT5 beta | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| BBS with 0.09% sodium azide; pH 8.2 | |
| P51692 | |
| Stat5b | |
| Peptide representing a region between residue 737 and the C-terminus (residue 787) of Human Signal Transducer and Activator of Transcription 5B. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| BLR146J | |
| Unconjugated | |
| Stat5b | |
| LOC101122961; mammary gland factor STAT5B; Signal transducer and activator of transcription 5a; signal transducer and activator of transcription 5B; STA5B; STAT; STAT5; STAT5B; transcription factor STAT5B | |
| Rabbit | |
| purified | |
| RUO | |
| 6777 | |
| 4°C | |
| Liquid |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering