missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT5A Antibody (251610), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB21741G
This item is not returnable.
View return policy
Description
STAT5A Monoclonal antibody specifically detects STAT5A in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| STAT5A | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 251610 | |
| Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| MGF, signal transducer and activator of transcription 5A, STAT5 | |
| Human STAT5a synthetic peptide, SLDSRLSPPAGLFTSARGSLS, Accession # NP_003143 | |
| 0.1 mL | |
| Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| 6776 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction