missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT5A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 302.00 - € 498.00
Specifications
| Antigen | STAT5A |
|---|---|
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18689395
|
Novus Biologicals
NBP2-46669-25ul |
25 μL |
€ 302.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18616475
|
Novus Biologicals
NBP2-46669 |
0.1 mL |
€ 498.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
STAT5A Polyclonal antibody specifically detects STAT5A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| STAT5A | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Signal Transduction, Transcription Factors and Regulators | |
| PBS (pH 7.2), 40% Glycerol | |
| MGF, signal transducer and activator of transcription 5A, STAT5 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: EGSLDVLQSWCEKLAEIIWQNRQQIRRAEHLCQQLPIPGPVEEMLAEVNATITDIISALVTSTFIIEKQPPQVL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| P42229 | |
| 6776 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title