missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT5A Antibody (OTI9F7), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74392APC
This item is not returnable.
View return policy
Description
STAT5A Monoclonal specifically detects STAT5A in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry.
Specifications
| STAT5A | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| STAT5A | |
| Full length recombinant protein of human STAT5A (NP_003143) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG1 |
| Flow Cytometry | |
| OTI9F7 | |
| Flow Cytometry | |
| MGF, signal transducer and activator of transcription 5A, STAT5 | |
| Mouse | |
| Affinity Purified | |
| Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| 6776.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction