missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT5A Antibody (OTI9F7), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74392H
This item is not returnable.
View return policy
Description
STAT5A Monoclonal specifically detects STAT5A in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| STAT5A | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| STAT5A | |
| Full length recombinant protein of human STAT5A (NP_003143) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI9F7 | |
| Western Blot, Immunohistochemistry | |
| MGF, signal transducer and activator of transcription 5A, STAT5 | |
| Mouse | |
| Affinity Purified | |
| Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| 6776.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction