missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT5a/b Antibody (SR2282), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21692-100ul
This item is not returnable.
View return policy
Description
STAT5a/b Monoclonal antibody specifically detects STAT5a/b in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Specifications
| STAT5a/b | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| SR2282 | |
| Western Blot 1:1000-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| MGF, STAT5, STAT5A signal transducer and activator of transcription 5A | |
| A synthesized peptide derived from human STAT5a/b (Uniprot #: P42229, P51692 ) | |
| 100 μg | |
| Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| 6776 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction