missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT5A Monoclonal Antibody (OTI11A9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502740S
This item is not returnable.
View return policy
Description
STAT5A Monoclonal antibody specifically detects STAT5A in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| STAT5A | |
| Monoclonal | |
| 0.86 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P42229 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6776 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI11A9 | |
| Unconjugated | |
| STAT5A | |
| AA959963; mammary gland factor; mammary gland factor STAT5A; mammary gland factor; transcription factor; Mgf; Mpf; signal transducer and activator of transcription 5; signal transducer and activator of transcription 5a; signal transducer and activator of transcription factor 5A; STAT; STAT5; STAT5A; STAT5A, Mammary Gland Factor; STAT5A/MGF; STAT5B; STATA5 | |
| Full length human recombinant protein of human STAT5A produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction