missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT6 Antibody (177C322.1), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-25241IR
This item is not returnable.
View return policy
Description
STAT6 Monoclonal specifically detects STAT6 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| STAT6 | |
| Monoclonal | |
| DyLight 755 | |
| NP_003144 | |
| STAT6 | |
| A portion of amino acids 600-650 of human Signal Transducer and Activator of Transcription 6 was used as the immunogen for the STAT6 Antibody (177C322.1). | |
| 0.1 mL | |
| Cancer, Cell Biology, Immune System Diseases, Immunology, Inflammation, Signal Transduction | |
| 6778 | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 177C322.1 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| D12S1644, EC 2.4.1.227, EC 2.7.7.6, IL-4 Stat, IL-4-STAT, signal transducer and activator of transcription 6, signal transducer and activator of transcription 6, interleukin-4 induced, STAT, interleukin-4 induced, STAT, interleukin4-induced, STAT6B, STAT6C, transcription factor IL-4 STAT | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This product reacts with Human and Mouse. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction