missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT6 Antibody (STAT6/7774R), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20997AF405
This item is not returnable.
View return policy
Description
STAT6 Monoclonal antibody specifically detects STAT6 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| STAT6 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| D12S1644, EC 2.4.1.227, EC 2.7.7.6, IL-4 Stat, IL-4-STAT, signal transducer and activator of transcription 6, signal transducer and activator of transcription 6, interleukin-4 induced, STAT, interleukin-4 induced, STAT, interleukin4-induced, STAT6B, STAT6C, transcription factor IL-4 STAT | |
| Recombinant fragment (around aa647-847) of human STAT6 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Apoptosis, Asthma, Cancer, Cell Biology, Epitope Tags, Immune System Diseases, Immunology, Inflammation, Phospho Specific, Signal Transduction | |
| 6778 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| STAT6/7774R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction