Learn More
STAT6, p Tyr641 Antibody (1248D), mFluor Violet 500 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals FAB3717MFV500
Product Code. 18539567
  • € 663.00 / 0.10mL
Description
STAT6 Monoclonal antibody specifically detects STAT6 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
STAT6
Monoclonal
mFluor Violet 500 SE
50mM Sodium Borate
Rabbit
0.1 mL
Apoptosis, Asthma, Cancer, Cell Biology, Epitope Tags, Immune System Diseases, Immunology, Inflammation, Phospho Specific, Signal Transduction
6778
Store at 4C in the dark.
Western Blot, Flow Cytometry, Immunocytochemistry
1248D
Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry
D12S1644, EC 2.4.1.227, EC 2.7.7.6, IL-4 Stat, IL-4-STAT, signal transducer and activator of transcription 6, signal transducer and activator of transcription 6, interleukin-4 induced, STAT, interleukin-4 induced, STAT, interleukin4-induced, STAT6B, STAT6C, transcription factor IL-4 STAT
Phosphopeptide containing human STAT6 Y641 site., Accession # P42226