Learn More
Invitrogen™ STAT6 Recombinant Rabbit Monoclonal Antibody (2F10)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560675
This item is not returnable.
View return policy
Description
STAT6 Recombinant Monoclonal Antibody for Western Blot, IHC (P), ELISA
STAT6 protein is a transcription factor activated by cytokines, particularly interleukin-4 and IL13. (Stat6-/-) were found to be deficient in IL-4-mediated functions including Th2 helper T-cell differentiation, expression of cell surface markers, T-cell proliferation, immunoglobulin class switching to IgE, and partial loss of IL-4 mediated proliferation. STAT6 mRNA has been detected in peripheral blood lymphocytes, colon, intestine, ovary, prostate, thymus, spleen, kidney, liver, lung and placenta. STAT6 is critically involved in Th2 immune response. STAT6 is crucial for IL-4-mediated biological responses, and naturally in two isoforms: STAT6a and STAT6c.
Specifications
| STAT6 | |
| Recombinant Monoclonal | |
| 0.6 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P42226, P52633 | |
| STAT6 | |
| A synthesized peptide derived from human STAT6. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2F10 | |
| Unconjugated | |
| STAT6 | |
| 12S1644; D12S1644; IL4 STAT; IL-4 Stat; IL-4-STAT; signal transducer and activator of transcription 6; signal transducer and activator of transcription 6, interleukin-4 induced; signal transducer and transcription activator 6; STAT; STAT, interleukin4-induced; Stat6; STAT-6; STAT6B; STAT6C; transcription factor IL-4 STAT | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20852, 6778 | |
| -20°C or -80°C if preferred | |
| Liquid |