missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STATH Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560755
This item is not returnable.
View return policy
Description
Immunogen sequence: LRRIGRFGYG YGPYQPVPEQ PLYPQPYQPQ YQQYTF Highest antigen sequence identity to the following orthologs: Mouse - 42%, Rat - 41%.
STATH is a salivary protein that stabilizes saliva supersaturated with calcium salts by inhibiting the precipitation of calcium phosphate salts. It also modulates hydroxyapatite crystal formation on the tooth surface.
Specifications
| STATH | |
| Polyclonal | |
| Unconjugated | |
| STATH | |
| STATH; Statherin; STR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6779 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P02808 | |
| STATH | |
| Recombinant protein corresponding to Human STATH. Recombinant protein control fragment (Product #RP-96641). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction