missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAU2, Mouse anti-Human, Clone: 3E9, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00027067-M09.100ug
This item is not returnable.
View return policy
Description
Sequence: LQINQMFSVQLSLGEQTWESEGSSIKKAQQAVANKALTESTLPKPVQKPPKSNVNNNPGSITPTVELNGLAMKRGEPAIYRPLDPKPFPSpecifications
| STAU2 | |
| Monoclonal | |
| Unconjugated | |
| staufen, RNA binding protein, homolog 2 (Drosophila) | |
| 39K2/39K3/DKFZp781K0371/MGC119606 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3E9 | |
| In 1x PBS, pH 7.4 | |
| NM_014393 | |
| STAU2 | |
| STAU2 (NP_055208, 2 a.a. ∼ 90 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 27067 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction