missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STAU2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582670
This item is not returnable.
View return policy
Description
Immunogen sequence: LQINQMFSVQ LSLGEQTWES EGSSIKKAQQ AVANKALTES TLPKPVQKPP KSNVNNNPGS ITPTVELNGL AMKRGEPAIY RPLDPKPFPN YRANYNFRGM YNQRYHCPVP KIFYVQLTVG NNEFFGEGKT RQAARHNAAM KALQ Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 56%.
Staufen homolog 2 is a member of the family of double-stranded RNA (dsRNA)-binding proteins involved in the transport and/or localization of mRNAs to different subcellular compartments and/or organelles. These proteins are characterized by the presence of multiple dsRNA-binding domains which are required to bind RNAs having double-stranded secondary structures. Staufen homolog 2 shares 48. 5% and 59. 9% similarity with drosophila and human staufen, respectively. The exact function of Staufen homolog 2 is not known, but since it contains 3 copies of conserved dsRNA binding domain, it could be involved in double-stranded RNA binding events. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
| STAU2 | |
| Polyclonal | |
| Unconjugated | |
| Stau2 | |
| 39K2; 39K3; Double-stranded RNA-binding protein Staufen homolog 2; r-staufen protein; Stau2; staufen (RNA binding protein) homolog 2 (Drosophila); staufen 2; staufen double-stranded RNA binding protein 2; staufen homolog 2; staufen, RNA binding protein, homolog 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 171500, 27067 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q68SB1, Q9NUL3 | |
| Stau2 | |
| Recombinant protein corresponding to Human STAU2. Recombinant protein control fragment (Product #RP-102290). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction