missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAU2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15998-1-AP-150UL
This item is not returnable.
View return policy
Description
STAU2 (Staufen homolog 2) is a member of the family of double-stranded RNA (dsRNA)-binding proteins involved in the transport and/or localization of mRNAs to different subcellular compartments and/or organelles. These proteins are characterized by the presence of multiple dsRNA-binding domains which are required to bind RNAs having double-stranded secondary structures. Staufen homolog 2 shares 48.5% and 59.9% similarity with drosophila and human staufen, respectively. The exact function of Staufen homolog 2 is not known, but since it contains 3 copies of conserved dsRNA binding domain, it could be involved in double-stranded RNA binding events. Several transcript variants encoding different isoforms have been found for this gene.Specifications
| STAU2 | |
| Polyclonal | |
| Unconjugated | |
| STAU2 | |
| 39K2, 39K3, STAU2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 171500, 27067, 29819 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q68SB1, Q8CJ67, Q9NUL3 | |
| STAU2 | |
| STAU2 Fusion Protein Ag8482 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction