Learn More
Staufen Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand:  Novus Biologicals NBP1-33202
Product Code. 18718163
  • € 602.00 / 0.10mL
Description
Staufen Polyclonal specifically detects Staufen in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot 1:500-1:3000, Immunoprecipitation 1:500-1:3000
63 kDa