Learn More
Staufen Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-33202
This item is not returnable.
View return policy
Description
Staufen Polyclonal specifically detects Staufen in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| Western Blot 1:500-1:3000, Immunoprecipitation 1:500-1:3000 | |
| 63 kDa |