missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STBD1 Monoclonal Antibody (OTI6G5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812280
This item is not returnable.
View return policy
Description
May have the capability to bind to carbohydrates.Specifications
| STBD1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O95210 | |
| STBD1 | |
| Human recombit protein fragment corresponding to amino acids 24-358 of human STBD1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI6G5 | |
| Unconjugated | |
| STBD1 | |
| GENEX3414, GENX-3414 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8987 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction