missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STCH Mouse anti-Human, Clone: 2F6, Novus Biologicals™
Mouse Monoclonal Antibody
€ 304.00 - € 453.00
Specifications
| Antigen | STCH |
|---|---|
| Clone | 2F6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18231572
|
Novus Biologicals
NBP2-60243-50UL |
50 μL |
€ 304.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18264163
|
Novus Biologicals
NBP2-60243-100UL |
100 μL |
€ 453.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
STCH Monoclonal specifically detects STCH in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| STCH | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 6782 | |
| Recombinant human HSPA13 (23-471aa) purified from E. coli | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 2F6 | |
| Western Blot 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| Human | |
| heat shock 70 kDa protein 13, heat shock protein 70kDa family, member 13, microsomal stress 70 protein ATPase core, Microsomal stress-70 protein ATPase core, STCHStress-70 protein chaperone microsome-associated 60 kDa protein, stress 70 protein chaperone, microsome-associated, 60kD, stress 70 protein chaperone, microsome-associated, 60kDa | |
| HSPA13 | |
| IgG1 κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title