missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEAP1 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76821H
This item is not returnable.
View return policy
Description
STEAP1 Polyclonal antibody specifically detects STEAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| STEAP1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| EC 1.16.1.-, PRSS24MGC19484, six transmembrane epithelial antigen of the prostate, six transmembrane epithelial antigen of the prostate 1, Six-transmembrane epithelial antigen of prostate 1, STEAPmetalloreductase STEAP1 | |
| Antibody was raised against a 18 amino acid synthetic peptide from near the carboxy terminus of human STEAP1. The immunogen is located within the last 50 amino acids of STEAP1. Amino Acid Squence: CLRKKILKIRHGWEDVTK | |
| 0.1 mL | |
| Cancer | |
| 26872 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction