missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEAP1 Antibody (J2D2), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-07094AF488
This item is not returnable.
View return policy
Description
STEAP1 Monoclonal specifically detects STEAP1 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| STEAP1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| NP_036581 | |
| STEAP1 | |
| Recombinant human STEAP1 (1-70aa) purified from E. coli (NP_036581). | |
| 0.1 mL | |
| Cancer | |
| 26872 | |
| Human, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| J2D2 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| EC 1.16.1.-, PRSS24MGC19484, six transmembrane epithelial antigen of the prostate, six transmembrane epithelial antigen of the prostate 1, Six-transmembrane epithelial antigen of prostate 1, STEAPmetalloreductase STEAP1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| STEAP1 (J2D2) | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction