missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STEAP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534225
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human (100%) Gorilla, Monkey, Marmoset (93%) Gibbon, Mouse, Bat, Horse (87%) Hamster, Elephant, Panda, Rabbit, Pig, Xenopus (80%).
The six-transmembrane epithelial antigen of prostate 4 (STEAP4) is a member of a family of metalloreductases identified as cell-surface antigens in prostate tissue. Similar to two other members of the STEAP family (STEAP 2 and STEAP3), STEAP4 promotes both iron and copper reduction. STEAP4 is highly expressed in placenta, lung, heart and prostate tissues, and overexpressed in prostate cancer cells compared to normal prostate cells. Overexpression of STEAP4 in prostate cells significantly increases cell growth and colony formation, suggesting STEAP4 may have a role in cell proliferation and prostate cancer progression.
Specifications
| STEAP4 | |
| Polyclonal | |
| Unconjugated | |
| STEAP4 | |
| 1110021O17Rik; Ab1-219; AI481214; DKFZp666D049; dudulin 4; dudulin-4; FLJ23153; Metalloreductase STEAP4; six transmembrane prostate protein 2; six-transmembrane epithelial antigen of prostate 4; SixTransMembrane protein of prostate 2; STAMP2; STEAP family member 4; STEAP4; STEAP4 metalloreductase; TIARP; Tnfa-induced adipose-related protein; Tnfaip9; Tumor necrosis factor, alpha-induced protein 9; tumor necrosis factor-alpha-induced adipose-related protein; tumor necrosis-alpha-induced adipose-related protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 79689 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q687X5 | |
| STEAP4 | |
| Synthetic 15 amino acid peptide from N-Terminus of human STEAP4. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction