missing translation for 'onlineSavingsMsg'
Learn More

Stearidonic Acid Ethyl Ester, TRC

Product Code. 30411942
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411942 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30411942 Supplier Toronto Research Chemicals Supplier No. TRCS68646010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Stearidonic acid ethyl ester
CAS 119798-44-6
Molecular Formula C20 H32 O2
Quantity 10 mg
Synonym (6Z,9Z,12Z,15Z)-6,9,12,15-Octadecatetraenoic Acid Ethyl Ester, (all-Z)- 6,9,12,15-Octadecatetraenoic Acid Ethyl Ester Ethyl Stearidonate, Moroctic Acid Ethyl Ester
InChI Formula InChI=1S/C20H32O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-4-2/h5-6,8-9,11-12,14-15H,3-4,7,10,13,16-19H2,1-2H3/b6-5-,9-8-,12-11-,15-14-
SMILES CCOC(=O)CCCC\C=C/C\C=C/C\C=C/C\C=C/CC
IUPAC Name ethyl (6Z,9Z,12Z,15Z)-octadeca-6,9,12,15-tetraenoate
Molecular Weight (g/mol) 304.47
Formula Weight 304.2402
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.