missing translation for 'onlineSavingsMsg'
Learn More

Stearyl Palmitate, TRC

Product Code. 30135033
Change view
Click to view available options
Quantity:
2.5 g
25 g
Unit Size:
2.5g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135033 25 g 25g
30003842 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135033 Supplier Toronto Research Chemicals Supplier No. TRCS68659025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2598-99-4
Molecular Formula C34H68O2
Molecular Weight (g/mol) 508.9
Synonym Hexadecanoic Acid Octadecyl Ester, Palmitic Acid Octadecyl Ester, Octadecyl Hexadecanoate, Octadecyl Palmitate, Purester 34;
IUPAC Name octadecyl hexadecanoate
SMILES CCCCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC

Specifications

Quantity 25 g
InChI Formula InChI=1S/C34H68O2/c1-3-5-7-9-11-13-15-17-18-19-21-23-25-27-29-31-33-36-34(35)32-30-28-26-24-22-20-16-14-12-10-8-6-4-2/h3-33H2,1-2H3
Formula Weight 508.522
Chemical Name or Material Stearyl Palmitate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.