Learn More
Usbeck Laborgeraete™ Steel/Ceramic Wire Cloths
Product Code. 10732702
Change view
Click to view available options
Length (Metric):
120 mm
160 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
10732702 160 mm Each
10372901 120 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10732702 Supplier Usbeck Laborgeraete™ Supplier No. 5082
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Ceramic Center Ceramic Center
Dimensions (L x W) 160 x 160 mm
Edges Folded
Quantity 1 Each
Shape Square
Length (Metric) 160 mm
Material Steel, Ceramic
Type Wire Cloth
Width (Metric) 160 mm