Learn More
RETSCH Steel O-ring
Shop All Retsch Products
Click to view available options
For Use With (Equipment):
Vibratory Disc Mill RS 200 250 mL Grinding Set
Vibratory Disc Mill RS 200 50 mL Grinding Set
Unit Size:
Each
Specifications
Specifications
| Product Type | O-ring |
| For Use With (Equipment) | Vibratory Disc Mill RS 200 50 mL Grinding Set |
| Material | Steel |
| Packaging Quantity | 1 Ea. |