Learn More
RETSCH Steel Spare Wearing Plates
Product Code. 10014881 Shop All Retsch Products
Click to view available options
For Use With (Equipment):
For BB 100 Jaw Crusher
For BB 200 Jaw Crusher
For BB 300 Jaw Crusher
Unit Size:
Each
Product Code. 10014881
Brand: RETSCH 22.711.0006
Only null left
Add to basket
Add to basket
Spare Wearing Plates
  • Accessory
Specifications
Product Type Spare Wearing Plate
For Use With (Equipment) For BB 200 Jaw Crusher
Material Steel