Learn More
Bochem™ Steel Zincked Standard Retort Clamps with Cork
Brand:  Bochem™ 5481
Additional Details : Weight : 0.41400kg
 View more versions of this product
Product Code. 10278540
  • € 31.55 / Each
Estimated Shipment: 21-05-2024
to see stock.
Specifications
1 Ea.
Zinc-plated Steel
180 mm
Stand Clamp
150 mm