missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies STEMdiff™ Neural Progenitor Medium

Product Code. 16803100
Encompass
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16803100 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16803100 Supplier STEMCELL Technologies Supplier No. 05833

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Medium for maintenance and expansion of neural progenitor cells derived from human ES and iPS cells

  • STEMdiff™ Neural Progenitor Medium is a defined and serum-free medium for the expansion of neural progenitor cells (NPCs) derived from human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells using STEMdiff™ Neural Induction Medium (Catalog #05835)
  • NPCs cultured in this medium can be expanded 3-5 fold per passage, and cultured for at least 10 passages, with minimal spontaneous neuronal differentiation

Specifications

Cell Type Neural Cells, PSC-Derived, Neural Stem and Progenitor Cells, Pluripotent Stem Cells
For Use With (Application) Cell Culture, Expansion
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.