missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies StemSpan™ Myeloid Expansion Supplement (100X)
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
- StemSpan™ Myeloid Expansion Supplement (100X) contains a combination of recombinant human cytokines formulated to selectively promote the expansion and differentiation of human myeloid progenitor cells, generating large numbers of CD15+ granulocytes from CD34+ cells isolated from human cord blood (CB) or bone marrow (BM) samples
- StemSpan™ Myeloid Expansion Supplement is intended for use in combination with any of the following StemSpan™ media: StemSpan™ SFEM (Catalog #09600)
- StemSpan™ SFEM II (Catalog #09605)
- StemSpan™-XF (Catalog #100-0073)
- StemSpan™-ACF Without Phenol Red (Catalog #100-0130)When added to serum-free medium, StemSpan™ Myeloid Expansion Supplement typically promotes the production of thousands of CD15+ granulocytes per input CD34+ cell in 14-day liquid cultures initiated with CD34+ human CB cells
- See data tab for more details
Specifications
Specifications
| Cell Type | Hematopoietic Stem and Progenitor Cells, Myeloid Cells |
| For Use With (Application) | Cell Culture, Differentiation, Expansion |
| Quantity | 1 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction