missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEP Antibody (23E5) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB300-202JF669
This item is not returnable.
View return policy
Description
STEP Monoclonal antibody specifically detects STEP in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Immunocytochemistry/ Immunofluorescence
Specifications
| STEP | |
| Monoclonal | |
| Janelia Fluor 669 | |
| EC 3.1.3.48, FLJ14427, Neural-specific protein-tyrosine phosphatase, protein tyrosine phosphatase, non-receptor type 5 (striatum-enriched), protein-tyrosine phosphatase striatum-enriched, STEPPTPSTEP, Striatum-enriched protein-tyrosine phosphatase, tyrosine-protein phosphatase non-receptor type 5 | |
| A synthetic peptide of rat STEP46 (amino acids 57-74: MCTPGCNEEGFGYLVSPR) | |
| 0.1 mL | |
| Alzheimers Research, Cancer, Cellular Markers, Hypoxia, Neurodegeneration, Neuroscience, Protein Phosphatase, Signal Transduction | |
| 84867 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunocytochemistry/Immunofluorescence | |
| 23E5 | |
| 50mM Sodium Borate | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction