missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ STEP Polyclonal Antibody

Product Code. 17159603
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17159603 100 μL 100µL
17138933 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17159603 Supplier Invitrogen™ Supplier No. PA5111688

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: EHTPIIVMIT NIEEMNEKCT EYWPEEQVAY DGVEITVQKV IHTEDYRLRL ISLKSGTEER GLKHY Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 91%.

Reversible phosphorylation of tyrosine residues is a key regulatory mechanism for numerous cellular events such as proliferation, differentiation, gene expression and migration. Abnormalities in tyrosine phosphorylation play a role in the pathogenesis of numerous inherited or acquired human diseases from cancer to immune deficiencies. There are at least 107 genes coding for PTPs (protein tyrosine phosphatases) in the human genome. All known PTPs contain a highly conserved 12 amino acid catalytic domain and are further distinguished on the basis of additional structural motifs. The receptor-like PTPs exhibit an extracellular domain, a single transmembrane and one or two intracellular phosphatase domains. The nonreceptor cytoplasmic PTPs contain a single phosphatase domain and additional amino acid sequences that are homologous to motifs found in other classes of proteins. Protein tyrosine phosphatases PTPN5 (STEP), PTPRR and PTPN7 comprise a family of phosphatases that specifically inactivate MAPKs (mitogen-activated protein kinases). There are multiple forms of STEP in the adult rat brain which show differential enrichment in brain regions implicated in aspects of cognitive, affective, and motor behaviors. Some of the STEP isoforms are generated through alternative splicing of a single STEP gene and each has unique intracellular targets and functions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STEP
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PTPN5
Gene Accession No. P54829
Gene Alias FLJ14427; Neural-specific protein-tyrosine phosphatase; protein tyrosine phosphatase, non-receptor type 5; protein tyrosine phosphatase, non-receptor type 5 (striatum-enriched); protein-tyrosine phosphatase striatum-enriched; protein-tyrosine-phosphatase non-receptor 5; PTN5; PTP STEP; PTPN 5; PTPN5; PTPSTEP; STEP; STEP61; striatal-enriched protein tyrosine phosphatase; striatum-enriched protein-tyrosine phosphatase; Tyrosine-protein phosphatase non-receptor type 5
Gene Symbols PTPN5
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human STEP. Recombinant protein control fragment (Product #RP-107861).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84867
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.