Learn More
Sterigmatocystin, TRC
Product Code. 30006393
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006393 5 mg 5mg
30010488 1 mg 1mg
30005609 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30006393 Supplier Toronto Research Chemicals Supplier No. TRCS6867105MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10048-13-2
Molecular Formula C18 H12 O6
Molecular Weight (g/mol) 324.28
Synonym 7H-Furo[3',2':4,5]furo[2,3-c]xanthen-7-one, 3a,12c-dihydro-8-hydroxy-6-methoxy- (7CI,8CI), 7H-Furo[3',2':4,5]furo[2,3-c]xanthen-7-one, 3a,12c-dihydro-8-hydroxy-6-methoxy-, (3aR-cis)-, Sterigmatocystin (6CI), (3aR,12cS)-3a,12c-Dihydro-8-hydroxy-6-methoxy-7H-furo[3',2':4,5]furo[2,3-c]xanthen-7-one, NSC 201423, NSC 204985, Sterigmatocystine
SMILES COc1cc2OC3OC=CC3c2c4Oc5cccc(O)c5C(=O)c14
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C18H12O6/c1-21-11-7-12-13(8-5-6-22-18(8)24-12)17-15(11)16(20)14-9(19)3-2-4-10(14)23-17/h2-8,18-19H,1H3
Formula Weight 324.0634
Chemical Name or Material Sterigmatocystin