Learn More
Dostmann Electronic SterilDisk Data Logger with probe 50 mm 0..140°C
Brand:  Dostmann Electronic 5005-0803
SterilDisk Data Logger with probe 50 mm 0..140°C
Product Code. 17822535
  • ONLINE EXCLUSIVE PRICE
    € 904.05
    € 589.00 / Each
    Save € 315.05
    34% Off