missing translation for 'onlineSavingsMsg'
Learn More

Heinz Herenz Sterile Wide Neck Bottles for Water Sampling

Product Code. 30158477
Click to view available options
:
For Water Sampling
This item is not returnable. View return policy

Product Code. 30158477

Brand: Heinz Herenz 1131046

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sterile wide neck bottles for water sampling, 250 ml,. With 5 mg sodium thiosulfate.

  • Bottles for water sampling
  • 250 mL
  • PP,
  • Transparent
  • With 5 mg sodium thiosulfate
  • Sterile
  • Wide neck

Specifications

Capacity (Metric) 250 mL
Capacity (English) 0.055 gal.
Color Clear
Material Polypropylene
Sterility Sterile
Closure Included Yes
Disposable Reusable
Graduated Yes
Mouth Wide
Shape Square
Fit for Purpose Sterile
Product Type Bottle
Autoclavable Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.