Learn More
Bel-Art™ Sterileware™ Pharma Scoop
Shop All SP Scienceware Products
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
2500 mL
Unit Size:
Pack of 100
Pack of 15
Pack of 25
Pack of 50
Pack of 85
Description
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High impact polystyrene (HIPS) scoops heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10-6)
- Ready for one time use and disposal
- Lot stamped for accurate tracing
- Shipped with a Certificate of Processing for Sterilization
Specifications
Specifications
| Quantity | 100/Pk. |
| Sterility | Sterile |
| Autoclavable | Not Autoclavable |
| Capacity (English) | 2 oz. |
| Capacity (Metric) | 60 mL |
| Product Type | Scoop |
| Dimensions (L x W) | Bowl: 7.2 x 4.9 x 2.8 cm (2.875 x 2 x 1.125 in.) |
| Length (English) Handle | 4.25 in. |
| Length (Metric) Handle | 10.6 cm |
| Material | High-impact Polystyrene |