Learn More
Bel-Art™ Sterileware™ Powder Thief
Product Code. 16692824 Shop All SP Scienceware Products
Click to view available options
Capacity (Metric):
1 mL
5 mL
10 mL
20 mL
Length (Metric):
50 cm
100 cm
Unit Size:
Pack of 20
Product Code. 16692824
Brand: Bel-Art™ F378740010
Powder Thief is available in two lengths and four tip volumes and easily samples from targeted locations within bulk powders.
  • Used by pharmaceutical, food, cosmetic and chemical companies.
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High density polyethylene (HDPE) thief heat sealed in individual polyethylene bag
  • Gamma irradiated sterile (SAL 10-6)
  • Ready for one time use and disposal
  • Lot stamped for accurate tracing
  • Shipped with a Certificate of Processing for Sterilization
Specifications
Quantity 20/Pk.
Sterility Sterile
Autoclavable Not Autoclavable
Capacity (Metric) 10 mL
Product Type Powder Sampler
Length (English) 19.75 in.
Material HDPE
Length (Metric) 50 cm